LIF189457882 Life Chemicals Inc.

1-[3-(methylsulfanyl)phenyl]guanidine

  • MDL number MFCD16653187
  • Cas Number : 71198-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart