LIF189457908 Life Chemicals Inc.

3-(4-Aminophenyl)pyrazole

  • MDL number MFCD03094636
  • Cas Number : 89260-45-7
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart