LIF189457929 Life Chemicals Inc.

3-Methoxy-2-methyl-2H-indazole-6-carboxylic acid

  • MDL number MFCD11655600
  • Cas Number : 919106-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart