LIF189457936 Life Chemicals Inc.

3-[5-Bromo-2-(difluoromethoxy)phenyl]propanoic acid

  • MDL number MFCD11655595
  • Cas Number : 1086386-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart