LIF189457938 Life Chemicals Inc.

tert-Butyl 6-hydroxy-3,4-dihydro-2(1H)-isoquinolinecarboxylate

  • MDL number MFCD08275034
  • Cas Number : 158984-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart