LIF189457944 Life Chemicals Inc.

2-(Methylsulfanyl)-5-pyrimidinecarbaldehyde

  • MDL number MFCD03780729
  • Cas Number : 90905-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart