LIF189457954 Life Chemicals Inc.

3-[3-(4-fluorophenoxy)phenyl]propanoic acid

  • MDL number MFCD11655656
  • Cas Number : 1096770-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart