LIF189457971 Life Chemicals Inc.

2-bromo-4,5-dimethyl-1,3-thiazole

  • MDL number MFCD11501629
  • Cas Number : 29947-24-8
  • Molecular Weight : 192.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart