LIF189457982 Life Chemicals Inc.

2-(4-oxo-1,2,3-benzotriazin-3(4H)-yl)propanoic acid

  • MDL number MFCD07636865
  • Cas Number : 51672-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart