LIF189458261 Life Chemicals Inc.

[4-(5-pyridin-3-yl-1H-1,2,4-triazol-3-yl)phenyl]amine

  • MDL number MFCD11058002
  • Cas Number : 1018168-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart