LIF189458264 Life Chemicals Inc.

{4-[5-(2-furyl)-1H-1,2,4-triazol-3-yl]phenyl}amine

  • MDL number MFCD11987044
  • Cas Number : 1049032-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart