LIF189458306 Life Chemicals Inc.

5-(2-thienylmethyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09041624
  • Cas Number : 926254-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart