LIF189459103 Life Chemicals Inc.

6-nitroquinolin-8-ol

  • MDL number MFCD00466084
  • Cas Number : 16727-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart