LIF189572689 Life Chemicals Inc.

5-(acetylamino)-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11986937
  • Cas Number : 51649-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart