LIF189572690 Life Chemicals Inc.

N-(2,2,2-trifluoroethyl)pyridin-3-amine

  • MDL number MFCD11986938
  • Cas Number : 77262-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart