LIF189573275 Life Chemicals Inc.

2-methyl-5,6-dihydro-1,4-oxathiine-3-carboxylic acid

  • MDL number MFCD00666851
  • Cas Number : 6577-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart