LIF189573285 Life Chemicals Inc.

tert-butyl 4-({[(4-methylphenyl)sulfonyl]oxy}methyl)piperidine-1-carboxylate

  • MDL number MFCD05864740
  • Cas Number : 166815-96-9
  • Molecular Weight : 369.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart