LIF189573289 Life Chemicals Inc.

2-amino-2-(3-chlorophenyl)acetic acid

  • MDL number MFCD00049326
  • Cas Number : 7292-71-9
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart