LIF189573296 Life Chemicals Inc.

6-methoxy-3-methyl-1-benzofuran-2-carboxylic acid

  • MDL number MFCD00514053
  • Cas Number : 10410-29-4
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart