LIF189573300 Life Chemicals Inc.

(4-methyl-2-phenyl-1,3-thiazol-5-yl)acetic acid

  • MDL number MFCD01681553
  • Cas Number : 34272-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart