LIF196580346 Life Chemicals Inc.

1,8-naphthyridine-3,6-diamine

  • MDL number MFCD16652618
  • Cas Number : 1177337-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart