LIF196580425 Life Chemicals Inc.

6-bromo-N-methyl-1,3-benzothiazol-2-amine

  • MDL number MFCD13190227
  • Cas Number : 75104-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart