LIF238935445 Life Chemicals Inc.

2-Bromo-3-methyl-1,5-naphthyridine

  • MDL number MFCD12404061
  • Cas Number : 1204297-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart