LIF238935447 Life Chemicals Inc.

5,7-dimethyl-1,2,3,4-tetrahydro-1,8-naphthyridine

  • MDL number MFCD13705330
  • Cas Number : 65541-95-9
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart