LIF238936201 Life Chemicals Inc.

N-methylcyclopentanamine

  • MDL number MFCD00019277
  • Cas Number : 2439-56-7
  • Molecular Weight : 99.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-methylcyclopentanamine
Inchi Code 1S/C6H13N/c1-7-6-4-2-3-5-6/h6-7H,2-5H2,1H3
Inchi Key KKTBUCVHSCATGB-UHFFFAOYSA-N

Description

Purity 95%
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart