LIF238936351 Life Chemicals Inc.

1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride

  • MDL number MFCD07366322
  • Cas Number : 124788-36-9
  • Molecular Weight : 238.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart