LIF238940421 Life Chemicals Inc.

N-benzyl-4-chloro-2-nitroaniline

  • MDL number MFCD01051337
  • Cas Number : 10066-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart