LIF238940422 Life Chemicals Inc.

5-amino-2-methylbenzenesulfonamide

  • MDL number MFCD06681071
  • Molecular Weight : 186.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart