LIF242633469 Life Chemicals Inc.

2-amino-4-(4-phenoxyphenyl)-1H-pyrrole-3-carbonitrile

  • MDL number MFCD14584888
  • Cas Number : 1199589-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart