LIF242633600 Life Chemicals Inc.

3-(4-methoxyphenyl)-2-phenylpropanoic acid

  • MDL number MFCD00046532
  • Cas Number : 4314-68-5
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart