LIF242633631 Life Chemicals Inc.

[4-(2-methylphenoxy)phenyl]amine

  • MDL number MFCD02663364
  • Cas Number : 56705-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart