LIF242633653 Life Chemicals Inc.

(1-acetylpiperidin-4-yl)(5-fluoro-2-hydroxyphenyl)methanone

  • MDL number MFCD06409282
  • Cas Number : 84162-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart