LIF242633656 Life Chemicals Inc.

3-butoxy-6-hydroxy-9H-xanthen-9-one

  • MDL number MFCD14584950
  • Cas Number : 164660-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart