LIF242633668 Life Chemicals Inc.

4-(3-thienyl)benzoic acid

  • MDL number MFCD03783559
  • Cas Number : 29886-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart