LIF242633669 Life Chemicals Inc.

ethyl 3-[5-bromo-2-(difluoromethoxy)phenyl]propanoate

  • MDL number MFCD09028856
  • Cas Number : 1057674-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart