LIF242634146 Life Chemicals Inc.

1-(4-fluorophenyl)-5-oxo-4,5-dihydro-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD16631642
  • Cas Number : 1204297-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart