LIF242634533 Life Chemicals Inc.

2-(thiophen-2-yl)cyclopropane-1-carboxylic acid

  • MDL number MFCD11186844
  • Cas Number : 90111-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart