LIF245822523 Life Chemicals Inc.

(2-{[(tert-butoxycarbonyl)amino]methyl}-1H-benzimidazol-1-yl)acetic acid

  • MDL number MFCD16640725
  • Cas Number : 1207044-25-4
  • Molecular Weight : 305.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart