LIF258630729 Life Chemicals Inc.

8-methoxy-2,3-dihydro-1,4-benzodioxine-6-carbaldehyde

  • MDL number MFCD06342801
  • Cas Number : 75889-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart