LIF258630761 Life Chemicals Inc.

2-amino-4,5,6,7-tetrahydro-1,3-benzothiazole-4-carboxylic acid

  • MDL number MFCD11902149
  • Cas Number : 40440-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart