LIF267312841 Life Chemicals Inc.

[4-(4-fluorophenyl)-1H-pyrazol-1-yl]acetic acid

  • MDL number MFCD16653380
  • Cas Number : 1206997-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart