LIF267313605 Life Chemicals Inc.

4-(2-methoxyphenyl)tetrahydro-2H-pyran-4-carboxylic acid

  • MDL number MFCD11188908
  • Cas Number : 1039931-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart