LIF267321530 Life Chemicals Inc.

1-(chloroacetyl)-4-[5-(trifluoromethyl)pyridin-2-yl]piperazine

  • MDL number MFCD16653302
  • Cas Number : 1219827-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart