LIF267321726 Life Chemicals Inc.

5-(2,4-dimethyl-1,3-thiazol-5-yl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11054009
  • Cas Number : 31877-60-8
  • Molecular Weight : 196.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart