LIF296039781 Life Chemicals Inc.

[2-(pyridin-2-ylamino)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD16631675
  • Cas Number : 1248907-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart