LIF296039786 Life Chemicals Inc.

2-(chloromethyl)-5-[(3-methoxybenzyl)oxy]-4H-pyran-4-one

  • MDL number MFCD16631680
  • Cas Number : 1248907-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart