LIF296040441 Life Chemicals Inc.

2-(chloromethyl)-8-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD03988539
  • Cas Number : 87591-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart