LIF296040442 Life Chemicals Inc.

6-fluoro-1H-indazole-3-carboxylic acid

  • MDL number MFCD03840713
  • Cas Number : 129295-30-3
  • Molecular Weight : 180.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-fluoro-1H-indazole-3-carboxylic acid
Inchi Code 1S/C8H5FN2O2/c9-4-1-2-5-6(3-4)10-11-7(5)8(12)13/h1-3H,(H,10,11)(H,12,13)
Inchi Key KNPFUABTEGLQBG-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart