LIF296040475 Life Chemicals Inc.

5-[4-(trifluoromethoxy)phenyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09054680
  • Cas Number : 910442-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart