LIF296040626 Life Chemicals Inc.

4-(chloromethyl)-2-(4-chlorophenyl)-1,3-thiazole

  • MDL number MFCD00047057
  • Cas Number : 17969-22-1
  • Molecular Weight : 244.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart